Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Mouse monoclonal [2B 10A8] Anti-Rat IgG2b heavy chain (Alexa Fluor™ 488), 250 μg

Catalog No. 502600745
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-260-0745 250 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-260-0745 Supplier Abcam Supplier No. AB172334250UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Mouse Monoclonal antibody, suitable for IHC-Fr, Flow Cyt, with 1 citations.

  • Host Species: Mouse
  • Clonality: Monoclonal
  • Isotype: IgG2b
  • Applications: IHC-Fr, Flow Cyt
  • Species Reactivity: Rat
  • Storage Buffer: Preservative: 0.02% Sodium azide, Constituents: PBS, 23% Glycerol (glycerin, glycerine), 1% BSA
  • Form: Liquid
  • Purification: Affinity purification Immunogen

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Applications Flow Cytometry, Immunohistochemistry (Frozen)
Classification Monoclonal
Clone 2B 10A8
Conjugate Unconjugated
Formulation Preservative: 0.02% Sodium azide, Constituents: PBS, 23% Glycerol (glycerin, glycerine), 1% BSA
Gene Rat IgG2b
Host Species Mouse
Purification Method Affinity purification Immunogen
Quantity 250 μg
Regulatory Status RUO
Whole Molecule Yes
Primary or Secondary Primary
Packaging Vial
Test Specificity This product reacts with Rat.
Target Species Rat
Product Type Monoclonal Antibody
Form Liquid
Isotype IgG2b
Host abrv Ms
Shipping Condition Blue Ice
Reactivity abrv Rt
Target Molecule Rat IgG2b
Validated Application abrv IHC-Fr, Flow Cyt
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.