Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Mouse monoclonal [M-4] Anti-Chicken IgM mu chain, 500 ug

Catalog No. 502663619
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-266-3619 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-266-3619 Supplier Abcam Supplier No. AB99715500UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Mouse Monoclonal antibody, suitable for Flow Cyt, WB, ICC/IF, IP.

  • Host Species: Mouse
  • Clonality: Monoclonal
  • Isotype: IgM
  • Applications: Flow Cyt, WB, ICC/IF, IP
  • Species Reactivity: Chicken and Turkey
  • Storage Buffer: pH: 8.2,
  • Form: Liquid

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Applications Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone M-4
Conjugate Unconjugated
Formulation pH: 8.2
Gene Ig mu chain C region
Host Species Mouse
Quantity 500 μg
Regulatory Status RUO
Whole Molecule Yes
Primary or Secondary Primary
Packaging Vial
Test Specificity This product reacts with Chicken and Turkey.
Target Species Chicken, Turkey
Product Type Monoclonal Antibody
Form Liquid
Isotype IgM
Host abrv Ms
Shipping Condition Blue Ice
Reactivity abrv Ck
Validated Application abrv Flow Cyt, WB, ICC/IF, IP
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.