Learn More
Abcam Mouse monoclonal [SB62a] Anti-Rabbit IgG light chain (HRP), 500 uL
Description
- Host Species: Mouse
- Clonality: Monoclonal
- Isotype: IgG2b
- Applications: ELISA, IHC-P, WB
- Species Reactivity: Rabbit
- Storage Buffer: pH: 7.4, Constituents: PBS, 50% Glycerol (glycerin, glycerine)
- Form: Liquid
- Purification: Affinity purification Protein A
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Applications | ELISA, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Clone | SB62a |
| Conjugate | Unconjugated |
| Formulation | pH: 7.4, Constituents: PBS, 50% Glycerol (glycerin, glycerine) |
| Gene | Rabbit IgG |
| Host Species | Mouse |
| Purification Method | Affinity purification Protein A |
| Quantity | 500 μL |
| Regulatory Status | RUO |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.