Learn More
Abcam Mouse monoclonal [UHB] Anti-Human IgM mu chain, 250 ug
Description
- Host Species: Mouse
- Clonality: Monoclonal
- Isotype: IgG3
- Applications: ELISA, Flow Cyt
- Species Reactivity: Human
- Storage Buffer: pH: 8.2, Constituents: 100% Borate buffered saline
- Form: Liquid
- Purification: Affinity purification Protein A
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Applications | ELISA, Flow Cytometry |
| Classification | Monoclonal |
| Clone | UHB |
| Conjugate | Unconjugated |
| Formulation | pH: 8.2, Constituents: 100% Borate buffered saline |
| Gene | IGHM |
| Gene Symbols | IGHM |
| Host Species | Mouse |
| Purification Method | Affinity purification Protein A |
| Quantity | 250 μg |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.