Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Mouse monoclonal [UHB] Anti-Human IgM mu chain (Alkaline Phosphatase), 500 uL

Catalog No. 502708186
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-270-8186 500 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-270-8186 Supplier Abcam Supplier No. AB99743500UL
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Mouse monoclonal [UHB] Anti-Human IgM mu chain (Alkaline Phosphatase)

  • Host Species: Mouse
  • Clonality: Monoclonal
  • Isotype: IgG3
  • Applications: Dot, ELISA, ICC, IHC-Fr, WB
  • Species Reactivity: Human
  • Storage Buffer: Glycerol, Magnesium chloride, Tris
  • Form: Liquid
  • Purification: Affinity purification Protein A

Specifications

Applications Dot Blot, Western Blot, Immunocytochemistry, Immunohistochemistry (Frozen), ELISA
Classification Monoclonal
Clone UHB
Conjugate Unconjugated
Formulation pH: 8, Preservative: 0.1% Sodium azide, Constituents: 50% Glycerol (glycerin, glycerine), 0.605% Tris, 0.0095% Magnesium chloride
Gene Symbols IGHM
Host Species Mouse
Purification Method Affinity purification Protein A
Quantity 500 μL
Regulatory Status RUO
Whole Molecule Yes
Primary or Secondary Secondary
Packaging Vial
Test Specificity This product reacts with Human.
Target Species Human
Product Type Monoclonal Secondary Antibody
Form Liquid
Isotype IgG3
Host abrv Ms
Shipping Condition Blue Ice
Reactivity abrv Hu
Validated Application abrv Dot, WB, ICC, IHC-Fr, ELISA
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.