Learn More
Abcam Native Cow Osteocalcin protein, 100 ug
Description
- Applications: RIA, ELISA, Dot, SDS-PAGE
- Storage Buffer: pH: 7.4, Constituents: 50% Glycerol (glycerin, glycerine), 0.435% Sodium chloride, 0.121% Tris
- Form: Liquid
- Purification: Affinity purification
Specifications
Specifications
| Formulation | pH: 7.4, Constituents: 50% Glycerol (glycerin, glycerine), 0.435% Sodium chloride, 0.121% Tris |
| Quantity | 100 μg |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.