Learn More
Abcam PE / R-Phycoerythrin Conjugation Kit - Lightning-Link™, 600 ug
Specifications
Specifications
| Product Type | Secondary Antibody |
| Shipping Condition | Blue Ice |
| Conjugate | PE |
| Quantity | 600 μ g |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.