Learn More
Abcam Phalloidin-iFluor 488 Reagent, 300 Tests
Description
- Applications: ICC/IF, Fluorescence Microscopy
- Storage Buffer: pH: 7.3, Constituents: 99% Dimethylsulfoxide, 1% Fluorescent-Phalloidin Conjugate
- Form: Liquid
Specifications
Specifications
| Formulation | pH: 7.3, Constituents: 99% Dimethylsulfoxide, 1% Fluorescent-Phalloidin Conjugate |
| Product Type | Secondary Antibody |
| Shipping Condition | Blue Ice |
| Conjugate | Unconjugated |
| Quantity | 300 Tests |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.