Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Phalloidin-iFluor 647 Reagent, 300 Tests

Catalog No. NC1458286
Encompass_Preferred
Change view
Click to view available options
Quantity:
300 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NC1458286 300 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NC1458286 Supplier Abcam Supplier No. AB176759300TEST
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Antibody, suitable for ICC/IF, Fluorescence Microscopy, with over 100 citations.

  • Applications: ICC/IF, Fluorescence Microscopy
  • Storage Buffer: Constituents: 100% Fluorescent-Phalloidin Conjugate
  • Form: Solid

Specifications

Formulation Constituents: 100% Fluorescent-Phalloidin Conjugate
Product Type Secondary Antibody
Shipping Condition Ambient
Conjugate Unconjugated
Quantity 300 Tests
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.