Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Rat AKI Antibody Array - Membrane (7 Targets), 1X8MEMBRANES

Catalog No. 502673068
Encompass_Preferred
Change view
Click to view available options
Quantity:
8 Membranes
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-267-3068 8 Membranes
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-267-3068 Supplier Abcam Supplier No. AB1936541X8MEMBRANES
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Antibody, suitable for Multiplex Protein Detection.

  • Applications: Multiplex Protein Detection
  • Species Reactivity: Rat

Specifications

Conjugate Unconjugated
Gene Symbols Cst3
Quantity 8 Membranes
Regulatory Status RUO
Whole Molecule Yes
Primary or Secondary Secondary
Test Specificity This product reacts with Rat.
Target Species Rat
Product Type Secondary Antibody
Detection Method Luminescent
Shipping Condition Blue Ice
Reactivity abrv Rt
Validated Application abrv Multiplex Protein Detection
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.