Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Rat Cytokine Antibody Array (27 Targets) - Quantitative, 50 Tests

Catalog No. 502652521
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-265-2521 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-265-2521 Supplier Abcam Supplier No. AB19748350TEST
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

Antibody.

  • Species Reactivity: Rat

Specifications

Conjugate Unconjugated
Gene Symbols Ager
Quantity 50 Tests
Regulatory Status RUO
Whole Molecule Yes
Primary or Secondary Secondary
Test Specificity This product reacts with Rat.
Target Species Rat
Product Type Secondary Antibody
Detection Method Fluorescent
Shipping Condition Blue Ice
Reactivity abrv Rt
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.