Learn More
Abcam Rat monoclonal [11-26] Anti-Mouse IgD delta chain (FITC), 250 ug
Description
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Antigen | Ig delta chain C region |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | 11-26 |
| Conjugate | Unconjugated |
| Formulation | pH: 7.4, Preservative: 0.1% Sodium azide, Constituents: PBS |
| Gene | Ig delta chain C region secreted form |
| Host Species | Rat |
| Purification Method | Affinity purification Protein A |
| Quantity | 250 μg |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.