Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Rat monoclonal [23G3] Anti-Mouse IgE epsilon chain (HRP), 500 uL

Catalog No. 502653630
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-265-3630 500 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-265-3630 Supplier Abcam Supplier No. AB99574500UL
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Rat monoclonal [23G3] Anti-Mouse IgE epsilon chain (HRP)

  • Host Species: Rat
  • Clonality: Monoclonal
  • Isotype: IgG1
  • Applications: ELISA, ICC/IF
  • Species Reactivity: Mouse
  • Storage Buffer: PBS, Glycerol
  • Form: Liquid
  • Purification: Affinity purification Protein A

Specifications

Applications ELISA, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone 23G3
Conjugate Unconjugated
Formulation pH: 7.4, Constituents: PBS, 50% Glycerol (glycerin, glycerine)
Gene IGHE
Gene Symbols IGHE
Host Species Rat
Purification Method Affinity purification Protein A
Quantity 500 μL
Regulatory Status RUO
Whole Molecule Yes
Primary or Secondary Primary
Packaging Vial
Test Specificity This product reacts with Mouse.
Target Species Mouse
Product Type Monoclonal Antibody
Form Liquid
Format Vial
Isotype IgG1
Host abrv Rt
Shipping Condition Blue Ice
Reactivity abrv Ms
Target Molecule IGHE, Mouse
Validated Application abrv ELISA, ICC/IF
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.