Learn More
Abcam Rat monoclonal Anti-Mouse IgG2a heavy chain (HRP) [LO-MG2a-9], 100 ug
Description
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Applications | ELISA |
| Classification | Monoclonal |
| Clone | LO-MG2a-9 |
| Conjugate | Unconjugated |
| Formulation | Preservative: 0.01% Thimerosal (merthiolate), Constituents: PBS, 50% Glycerol (glycerin, glycerine) |
| Gene | Mouse IgG2a |
| Host Species | Rat |
| Purification Method | Affinity purification Protein G |
| Quantity | 100 μg |
| Regulatory Status | RUO |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.