Learn More
Abcam Rat monoclonal [LO-MM-9] Anti-Mouse IgM heavy chain (Biotin), 250 ug
Description
- Host Species: Rat
- Clonality: Monoclonal
- Isotype: IgG2a
- Applications: ELISA
- Species Reactivity: Mouse
- Storage Buffer: pH: 7.2, Preservative: 0.1% Sodium azide, Constituents: PBS
- Form: Liquid
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Applications | ELISA |
| Classification | Monoclonal |
| Clone | LO-MM-9 |
| Conjugate | Unconjugated |
| Formulation | pH: 7.2, Preservative: 0.1% Sodium azide, Constituents: PBS |
| Gene | Ighm |
| Gene Symbols | Ighm |
| Host Species | Rat |
| Quantity | 250 μg |
| Regulatory Status | RUO |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.