Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Rat monoclonal [SB74g] Anti-Mouse IgG2b gamma chain (Alexa Fluor™ 647), 250 ug

Catalog No. 502612920
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-261-2920 250 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-261-2920 Supplier Abcam Supplier No. AB172327250UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Rat Monoclonal antibody, suitable for Flow Cyt, ELISA, IHC-P, ICC/IF, with 4 citations.

  • Host Species: Rat
  • Clonality: Monoclonal
  • Isotype: IgG2b
  • Applications: Flow Cyt, ELISA, IHC-P, ICC/IF
  • Species Reactivity: Mouse
  • Storage Buffer: Preservative: 0.02% Sodium azide, Constituents: PBS, 23% Glycerol (glycerin, glycerine), 1% BSA
  • Form: Liquid

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Applications ELISA, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone SB74g
Conjugate Unconjugated
Formulation Preservative: 0.02% Sodium azide, Constituents: PBS, 23% Glycerol (glycerin, glycerine), 1% BSA
Gene Mouse IgG2b
Host Species Rat
Quantity 250 μg
Regulatory Status RUO
Whole Molecule Yes
Primary or Secondary Primary
Packaging Vial
Test Specificity This product reacts with Mouse.
Target Species Mouse
Product Type Monoclonal Antibody
Form Liquid
Isotype IgG2b
Host abrv Rt
Shipping Condition Blue Ice
Reactivity abrv Ms
Target Molecule Mouse IgG2b
Validated Application abrv Flow Cyt, ELISA, IHC-P, ICC/IF
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.