Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Rat monoclonal [SB77e] Anti-Mouse IgG1 H&L (FITC) preadsorbed, 250 ug

Catalog No. 502608795
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-260-8795 250 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-260-8795 Supplier Abcam Supplier No. AB133859250UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Rat Monoclonal antibody, suitable for ELISA, Flow Cyt, ICC/IF.

  • Host Species: Rat
  • Clonality: Monoclonal
  • Isotype: IgG2b
  • Applications: ELISA, Flow Cyt, ICC/IF
  • Species Reactivity: Mouse
  • Storage Buffer: pH: 7.4, Preservative: 0.1% Sodium azide, Constituents: PBS
  • Form: Liquid
  • Purification: Affinity purification

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Applications ELISA, Flow Cytometry, Immunocytochemistry/Immunofluorescence
Classification Monoclonal
Clone SB77e
Conjugate Unconjugated
Formulation pH: 7.4, Preservative: 0.1% Sodium azide, Constituents: PBS
Gene Ighg1
Gene Symbols Ighg1
Host Species Rat
Purification Method Affinity purification
Quantity 250 μg
Regulatory Status RUO
Whole Molecule Yes
Primary or Secondary Primary
Packaging Vial
Test Specificity This product reacts with Mouse.
Target Species Mouse
Product Type Monoclonal Antibody
Form Liquid
Isotype IgG2b
Host abrv Rt
Shipping Condition Blue Ice
Target Molecule Ighg1, Mouse
Validated Application abrv ELISA, Flow Cyt, ICC/IF
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.