Learn More
Abcam Sheep F(ab) Anti-Guinea pig IgG H&L (HRP), 500 uL
Description
- Host Species: Sheep
- Clonality: Polyclonal
- Isotype: IgG
- Applications: ELISA, ICC/IF, IHC-Fr, IHC-P, WB
- Species Reactivity: Guinea pig
- Storage Buffer: Constituents: 50% Glycerol (glycerin, glycerine), 49% Tris buffered saline
- Form: Liquid
- Purification: Affinity purification Immunogen
Specifications
Specifications
| Applications | ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Conjugate | HRP |
| Formulation | Constituents: 50% Glycerol (glycerin, glycerine), 49% Tris buffered saline |
| Host Species | Sheep |
| Purification Method | Affinity purification Immunogen |
| Quantity | 500 μL |
| Regulatory Status | RUO |
| Whole Molecule | Yes |
| Primary or Secondary | Secondary |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.