Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Total OXPHOS Rodent WB Antibody Cocktail, 300 ug

Catalog No. NC1912210
Encompass_Preferred
Change view
Click to view available options
Quantity:
300 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NC1912210 300 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NC1912210 Supplier Abcam Supplier No. AB110413300UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Antibody, suitable for WB, with over 1,000 citations.

  • Applications: WB
  • Species Reactivity: Mouse, Rat, Cow, Human and Cynomolgus monkey
  • Storage Buffer: Preservative: 0.02% Sodium azide, Constituents: 0.15% HEPES
  • Form: Liquid

Specifications

Applications Western Blot
Conjugate Unconjugated
Formulation Preservative: 0.02% Sodium azide, Constituents: 0.15% HEPES
Gene Symbols NDUFB8
Quantity 300 μg
Regulatory Status RUO
Whole Molecule Yes
Primary or Secondary Secondary
Test Specificity This product reacts with Mouse, Rat, Cow, Human and Cynomolgus monkey.
Target Species Mouse, Rat, Bovine, Human, Cynomolgus Monkey
Product Type Secondary Antibody
Form Liquid
Shipping Condition Blue Ice
Reactivity abrv Ms, Rt, Cow, Hu
Validated Application abrv WB
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.