Learn More
Abcam VHH Single Domain Anti-Rabbit IgG Fc (HRP), 250 uL
Description
- Clonality: Monoclonal
- Applications: IHC-P, WB, ELISA
- Species Reactivity: Rabbit
- Storage Buffer: pH: 7.4, Preservative: 0.1% Proclin 300 Solution, Constituents: Sodium phosphate, Sodium chloride, PBS, 30% Glycerol (glycerin, glycerine), 1% BSA
- Form: Liquid
- Purification: Affinity purification His Tag
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Applications | ELISA, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Clone | GD001 |
| Conjugate | Unconjugated |
| Formulation | pH: 7.4, Preservative: 0.1% Proclin 300 Solution, Constituents: Sodium phosphate, Sodium chloride, PBS, 30% Glycerol (glycerin, glycerine), 1% BSA |
| Gene | Rabbit IgG |
| Purification Method | Affinity purification His Tag |
| Quantity | 250 μL |
| Regulatory Status | RUO |
| Whole Molecule | Yes |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.