Learn More
Enzo Life Sciences P-glycoprotein (human) monoclonal antibody (C494) (1ml)
SDP
Supplier:  Enzo Life Sciences ALX801003C100
Encompass_Preferred
Clone: C494. Isotype: IgG2a. Immunogen: SDS-solubilized plasma membranes of an MDR (multidrug resistant) Chinese hamster ovary cell line and a human leukemic cell line. Species reactivity: Human. Specificity: Recognizes a gene-specific, internal cellular epitope present only on the human MDR1 isoform of P-glycoprotein. Applications: Flow Cytometry, ICC, IHC (FS), IHC (PS), IP, WB. Formulation: Liquid. In PBS containing BSA and 0.1% sodium azide.
Catalog No. 50-200-9681