Learn More
Enzo Life Sciences P-glycoprotein Recombinant monoclonal antibody (MRK16) (Mouse IgG1 kappa) (100µg)
SDP
Supplier:  Enzo Life Sciences ENZABS5580100
Encompass_Preferred
Clone: MRK16. Isotype: IgG1κ. Immunogen: Drug resistant K-562 cells. Species reactivity: Human. Specificity: Recognizes a discontinuous extracellular epitope on P-glycoprotein. Applications: Flow Cytometry, IF, IP. Purity: Protein A affinity purified. Formulation: Liquid. In PBS containing 0.02% Proclin 300.
Catalog No. 50-201-3084