Learn More
Aviva Systems Biology Corp ABCB4 Antibody - middle region (ARP41742_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP41742_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ABCB4. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human ABCB4. Alternate names: ABC21, GBD1, MDR2, MDR2/3, MDR3, PFIC-3, PGY3ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep
Catalog No. 50-014-57976