Learn More
Aviva Systems Biology Corp ABCC11 Antibody - middle region (ARP43719_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP43719_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ABCC11. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human ABCC11. Alternate names: EWWD, MRP8, WWApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Yeast
Catalog No. 50-014-58612