Learn More
Aviva Systems Biology Corp Abcc12 Antibody - middle region (ARP43708_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP43708_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Abcc12. It was validated on Western Blot. The immunogen is a synthetic peptide corresponding to a region of Mouse. Alternate names: 4930467B22Rik, MRP9ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-285-094