Learn More
Aviva Systems Biology Corp ABCC3 Antibody - middle region (ARP43645_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP43645_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ABCC3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human ABCC3. Alternate names: ABC31, EST90757, MLP2, MOAT-D, MRP3, cMOAT2ApplicationWB. Species reactivity: Dog, Guinea Pig, Horse, Human, Pig, Rat
Catalog No. 50-014-58597