Learn More
Aviva Systems Biology Corp ABCC9 Antibody - middle region (ARP43664_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP43664_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ABCC9. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human ABCC9. Alternate names: ABC37, CMD1O, FLJ36852, SUR2, ATFB12ApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-53372