Learn More
Aviva Systems Biology Corp ABCD2 Antibody - middle region (ARP43658_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP43658_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ABCD2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human ABCD2. Alternate names: ABC39, ALDL1, ALDR, ALDRP, hALDRApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Yeast, Zebrafish
Catalog No. 50-014-58601