Learn More
Aviva Systems Biology Corp ABCD2 Antibody - N-terminal region (ARP43657_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP43657_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ABCD2. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human ABCD2. Alternate names: ABC39, ALDL1, ALDR, ALDRP, hALDRApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-285-087