Learn More
Aviva Systems Biology Corp ABCD4 Antibody - C-terminal region (ARP43656_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP43656_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against ABCD4. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human ABCD4. Alternate names: ABC41, EST352188, P70R, P79R, PMP69, PXMP1LApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Yeast
Catalog No. 50-014-53371