Learn More
Aviva Systems Biology Corp ABCE1 Antibody - C-terminal region (ARP43633_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP43633_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ABCE1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human ABCE1. Alternate names: ABC38, OABP, RLI, RNASEL1, RNASELI, RNS4IApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-58593