Learn More
Aviva Systems Biology Corp Abcf1 Antibody - C-terminal region (ARP43631_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP43631_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Abcf1. It was validated on Western Blot. The immunogen is a synthetic peptide corresponding to a region of Rat. Alternate names: Abc50, Abcf1ApplicationIP, WB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-285-086