Learn More
Aviva Systems Biology Corp ABCF2 Antibody - N-terminal region (ARP43665_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP43665_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ABCF2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human ABCF2. Alternate names: ABC28, DKFZp586K1823, EST133090, HUSSY-18, M-ABC1, HUSSY18ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-58602