Learn More
Aviva Systems Biology Corp ABCF3 Antibody - N-terminal region (ARP43684_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP43684_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ABCF3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human ABCF3. Alternate names: EST201864, FLJ11198ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-58606