Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC ABCG2 protein, human (HEK293) | 10UG
SDP

Catalog No. 5000175897
Encompass_Preferred

ABCG2 Protein, Human (HEK293) is a recombinant human ATP-binding cassette transporter produced in HEK293 cells and provided as a detergent-containing, filtered protein solution for biochemical and transporter studies.

  • Human sequence, 655 amino acids.
  • Approximate molecular weight 72.5 kDa.
  • Endotoxin level <1 EU/μg.
  • Supplied in 40 mM HEPES (pH 7.5), 150 mM NaCl, 0.005% (w/v) GDN.
  • 0.22 μm filtered for sterility and clarity.
  • Store at -80°C and avoid repeated freeze-thaw cycles.
  • Suitable for transporter function and drug-resistance studies.

Catalog No. 50-001-75897 Supplier Medchemexpress LLC Supplier No. HYP70326610UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

ABCG2 Protein, Human (HEK293) is a recombinant human ATP-binding cassette transporter produced in HEK293 cells and provided as a detergent-containing, filtered protein solution for biochemical and transporter studies.

  • Human sequence, 655 amino acids.
  • Approximate molecular weight 72.5 kDa.
  • Endotoxin level <1 EU/μg.
  • Supplied in 40 mM HEPES (pH 7.5), 150 mM NaCl, 0.005% (w/v) GDN.
  • 0.22 μm filtered for sterility and clarity.
  • Store at -80°C and avoid repeated freeze-thaw cycles.
  • Suitable for transporter function and drug-resistance studies.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.