Learn More
Medchemexpress LLC Abd957 | 3007772-60-0 | 98.1% | 627.68 g/mol | C27H36F3N7O5S | 5 MG
SDP
Catalog No. 5000221701
Encompass_Preferred
ABD957 is a selective, covalent inhibitor of the ABHD17 family of depalmitoylases, reported to inhibit ABHD17B with an IC50 of 0.21 μM; it is supplied for research use with analytical data provided for confirmation.
  • Selective covalent inhibition of ABHD17 depalmitoylases with reported IC50 0.21 μM for ABHD17B.
  • Reported purity supports biochemical and cellular assays.
  • Characterized molecular weight and chemical formula for analytical verification.
  • Available in multiple small-scale quantities suitable for laboratory studies.
Catalog No. 50-002-21701 Supplier Medchemexpress LLC Supplier No. HY1421615MG
ABD957 is a selective, covalent inhibitor of the ABHD17 family of depalmitoylases, reported to inhibit ABHD17B with an IC50 of 0.21 μM; it is supplied for research use with analytical data provided for confirmation.
  • Selective covalent inhibition of ABHD17 depalmitoylases with reported IC50 0.21 μM for ABHD17B.
  • Reported purity supports biochemical and cellular assays.
  • Characterized molecular weight and chemical formula for analytical verification.
  • Available in multiple small-scale quantities suitable for laboratory studies.