Learn More
Medchemexpress LLC ABD957 | 3007772-60-0 | 98.14% | C27H36F3N7O5S | 100 MG
SDP
Catalog No. 5000355065
Encompass_Preferred
ABD957 is a potent and selective covalent inhibitor of the ABHD17 family of depalmitoylases, with an IC50 of 0.21 μM for ABHD17B. It can block N-Ras signaling and the growth of NRAS-mutant AML cells.
  • Blocks N-Ras signaling
  • Blocks the growth of NRAS-mutant AML cells
Catalog No. 50-003-55065 Supplier Medchemexpress LLC Supplier No. HY142161100MG
ABD957 is a potent and selective covalent inhibitor of the ABHD17 family of depalmitoylases, with an IC50 of 0.21 μM for ABHD17B. It can block N-Ras signaling and the growth of NRAS-mutant AML cells.
  • Blocks N-Ras signaling
  • Blocks the growth of NRAS-mutant AML cells