Learn More
Cole-Parmer Large Capacity Beaker with Handle, PP, 10L, 1 pack
Supplier:  Cole-Parmer 6100057
Encompass_Preferred
Abdos P50808 Large Capacity Beaker with Handle, Polypropylene, 10L, 1 pack, No need to mix multiple smaller volumesConstructed of durable polypropyleneAutoclavableBeaker features a 10,000 mL capacity with 1000 mL graduationsDesigned with handle for easy pouring
Catalog No. NC1249152