Learn More
Medchemexpress LLC HY-16297A 500mg , Abemaciclib CAS:1231929-97-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY16297A500MG
Encompass_Preferred
HY-16297A 500mg Abemaciclib CAS: 1231929-97-7 Abemaciclib (LY2835219) is a selective CDK4/6 inhibitor with IC50 values of 2 nM and 10 nM for CDK4 and CDK6, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9026