Learn More
Advanced Chemblocks Inc ABEMACICLIB
Supplier:  Advanced Chemblocks Inc O34190250MG
Encompass_Preferred
Advanced ChemBlocks Inc, Cat ID: O-34190, abemaciclib, CAS: 1231929-97-7, MDL: MFCD22665744, FW: 506.61, Formula: C27H32F2N8, IUPAC Name: n-(5-((4-ethylpiperazin-1-yl)methyl)pyridin-2-yl)-5-fluoro-4-(4-fluoro-1-isopropyl-2-methyl-1h-benzo[d]imidazol-6-yl)pyrimidin-2-amine
Catalog No. NC1887971
 Restricted Item