Learn More
Medchemexpress LLC Abemaciclib-d8 | 2088650-53-5 | 99.9% | 514.64 | 1 MG
SDP
Catalog No. 5000394510
Encompass_Preferred
Abemaciclib-d8 is the deuterium labeled Abemaciclib. Abemaciclib is a selective CDK4/6 inhibitor with IC50 values of 2 nM and 10 nM for CDK4 and CDK6, respectively. Stable heavy isotopes have been incorporated into drug molecules, largely as tracers for quantitation during the drug development process. Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes have potential to affect the pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-003-94510 Supplier Medchemexpress LLC Supplier No. HY16297AS1MG
Abemaciclib-d8 is the deuterium labeled Abemaciclib. Abemaciclib is a selective CDK4/6 inhibitor with IC50 values of 2 nM and 10 nM for CDK4 and CDK6, respectively. Stable heavy isotopes have been incorporated into drug molecules, largely as tracers for quantitation during the drug development process. Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes have potential to affect the pharmacokinetic and metabolic profiles of drugs.