Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Abemaciclib Impurity 1 | 1231930-33-8 | 99.4% | C11H12BrFN2 | 1 G
SDP

Catalog No. 5000434850
Encompass_Preferred

Abemaciclib Impurity 1 (6-Bromo-4-fluoro-1-isopropyl-2-methyl-1H-benzo[d]imidazole) is a drug intermediate for the synthesis of various active compounds. This product is for research use only and has not been fully validated for medical applications.

  • Target: Drug intermediate
  • Pathway: Others
  • Store powder at -20°C for 3 years or 4°C for 2 years
  • Store in solvent at -80°C for 6 months or -20°C for 1 month
  • For research use only

Catalog No. 50-004-34850 Supplier Medchemexpress LLC Supplier No. HY780661G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Abemaciclib Impurity 1 (6-Bromo-4-fluoro-1-isopropyl-2-methyl-1H-benzo[d]imidazole) is a drug intermediate for the synthesis of various active compounds. This product is for research use only and has not been fully validated for medical applications.

  • Target: Drug intermediate
  • Pathway: Others
  • Store powder at -20°C for 3 years or 4°C for 2 years
  • Store in solvent at -80°C for 6 months or -20°C for 1 month
  • For research use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.