Learn More
Medchemexpress LLC Abemaciclib Impurity 1 | 1231930-33-8 | C11H12BrFN2 | 50 G
SDP
Catalog No. 5000433195
Encompass_Preferred
Abemaciclib Impurity 1 (6-Bromo-4-fluoro-1-isopropyl-2-methyl-1H-benzo[d]imidazole) is a drug intermediate for the synthesis of various active compounds.
  • Keep container tightly sealed in a cool, well-ventilated area
  • Keep away from direct sunlight and sources of ignition
  • Recommended storage temperature for powder: -20°C for 3 years or 4°C for 2 years
  • Recommended storage temperature in solvent: -80°C for 6 months or -20°C for 1 month
  • Shipping at room temperature if less than 2 weeks
Catalog No. 50-004-33195 Supplier Medchemexpress LLC Supplier No. HY7806650G
Abemaciclib Impurity 1 (6-Bromo-4-fluoro-1-isopropyl-2-methyl-1H-benzo[d]imidazole) is a drug intermediate for the synthesis of various active compounds.
  • Keep container tightly sealed in a cool, well-ventilated area
  • Keep away from direct sunlight and sources of ignition
  • Recommended storage temperature for powder: -20°C for 3 years or 4°C for 2 years
  • Recommended storage temperature in solvent: -80°C for 6 months or -20°C for 1 month
  • Shipping at room temperature if less than 2 weeks