Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Abemaciclib (LY2835219) | 1231929-97-7 | MFCD22665744 | 99.9% | 506.6 g/mol | C27H32F2N8 | 10 MG
SDP

Catalog No. 5000288715
Encompass_Preferred

Analytical standard of Abemaciclib (LY2835219) provided for research and analytical applications. The material is intended as a reference standard for assay calibration, method validation, and LC-MS/HPLC analysis, with a high purity specification to support reproducible analytical results.

  • Intended for research and analytical applications as an analytical reference standard.
  • High purity suitable for LC-MS and HPLC assay calibration (manufacturer-listed purity ≈99.9%).
  • Useful for method validation, calibration, and quantitative analysis of abemaciclib.
  • Small pack sizes available for analytical workflows (e.g., 10 mg).
  • Chemical identifiers: CAS 1231929-97-7; molecular formula C27H32F2N8.

Catalog No. 50-002-88715 Supplier Medchemexpress LLC Supplier No. HY16297AR10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Analytical standard of Abemaciclib (LY2835219) provided for research and analytical applications. The material is intended as a reference standard for assay calibration, method validation, and LC-MS/HPLC analysis, with a high purity specification to support reproducible analytical results.

  • Intended for research and analytical applications as an analytical reference standard.
  • High purity suitable for LC-MS and HPLC assay calibration (manufacturer-listed purity ≈99.9%).
  • Useful for method validation, calibration, and quantitative analysis of abemaciclib.
  • Small pack sizes available for analytical workflows (e.g., 10 mg).
  • Chemical identifiers: CAS 1231929-97-7; molecular formula C27H32F2N8.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.