Learn More
Medchemexpress LLC Abemaciclib metabolite M18 | 2704316-81-2 | 98.01% | 10 MG
SDP
Catalog No. 5000338869
Encompass_Preferred
Abemaciclib metabolite M18 (LSN3106729) is a metabolite of Abemaciclib and acts as a CDK inhibitor with antitumor activity. It has been utilized in conjunction with a CRBN ligand to engineer a PROTAC CDK4/6 degrader. The primary mechanism of action for CDK4/6 inhibitors involves the inhibition of retinoblastoma (RB) protein phosphorylation, leading to the induction of cell cycle arrest.
  • Acts as a CDK inhibitor with antitumor activity.
  • Utilized in conjunction with a CRBN ligand to engineer a PROTAC CDK4/6 degrader.
  • Inhibits retinoblastoma (RB) protein phosphorylation.
  • Induces cell cycle arrest.
  • Promotes T-cell persistence and increases immunologic memory in mice that receive tumor-specific CD8+ T cells.
  • Appears as an off-white to light yellow solid.
Catalog No. 50-003-38869 Supplier Medchemexpress LLC Supplier No. HY126534A10MG
Abemaciclib metabolite M18 (LSN3106729) is a metabolite of Abemaciclib and acts as a CDK inhibitor with antitumor activity. It has been utilized in conjunction with a CRBN ligand to engineer a PROTAC CDK4/6 degrader. The primary mechanism of action for CDK4/6 inhibitors involves the inhibition of retinoblastoma (RB) protein phosphorylation, leading to the induction of cell cycle arrest.
  • Acts as a CDK inhibitor with antitumor activity.
  • Utilized in conjunction with a CRBN ligand to engineer a PROTAC CDK4/6 degrader.
  • Inhibits retinoblastoma (RB) protein phosphorylation.
  • Induces cell cycle arrest.
  • Promotes T-cell persistence and increases immunologic memory in mice that receive tumor-specific CD8+ T cells.
  • Appears as an off-white to light yellow solid.