Learn More
Medchemexpress LLC Abemaciclib metabolite M18 hydrochloride | 2704316-82-3 | 98.2% | 100 MG
SDP
Catalog No. 5000338887
Encompass_Preferred
Abemaciclib metabolite M18 hydrochloride (LSN3106729) is a metabolite of Abemaciclib with antitumor activity, acting as a CDK inhibitor. It has been utilized in the design of PROTAC CDK4/6 degraders.
  • Targets cyclin-dependent kinases (CDK4 and CDK6)
  • Exhibits antitumor activity
  • Soluble in DMSO up to 125 mg/mL for in vitro studies
  • Soluble in H2O up to 100 mg/mL for in vitro studies
  • Soluble in PBS up to 50 mg/mL for in vivo applications
  • Appears as an off-white to light yellow solid
Catalog No. 50-003-38887 Supplier Medchemexpress LLC Supplier No. HY126534A100MG
Abemaciclib metabolite M18 hydrochloride (LSN3106729) is a metabolite of Abemaciclib with antitumor activity, acting as a CDK inhibitor. It has been utilized in the design of PROTAC CDK4/6 degraders.
  • Targets cyclin-dependent kinases (CDK4 and CDK6)
  • Exhibits antitumor activity
  • Soluble in DMSO up to 125 mg/mL for in vitro studies
  • Soluble in H2O up to 100 mg/mL for in vitro studies
  • Soluble in PBS up to 50 mg/mL for in vivo applications
  • Appears as an off-white to light yellow solid