Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Abemaciclib metabolite M18 (hydrochloride) | 2704316-82-3 | 98.2% | 50 MG
SDP

Catalog No. 5000339675
Encompass_Preferred

Abemaciclib metabolite M18 hydrochloride is a metabolite of Abemaciclib and functions as a CDK inhibitor with antitumor activity. It has been used to design PROTAC CDK4/6 degraders. This product is for research use only, suitable for laboratory chemical applications and experienced personnel. It inhibits retinoblastoma (RB) protein phosphorylation, inducing cell cycle arrest, and can increase T-cell persistence and immunologic memory.

  • Solid appearance
  • Purity of 98.18%
  • Molecular weight: 531.00
  • Chemical formula: C25H29ClF2N8O
  • Stable under recommended storage conditions
  • Not classified as a hazardous substance or mixture
  • Acts as a CDK inhibitor
  • Exhibits antitumor activity
  • Inhibits retinoblastoma (RB) protein phosphorylation
  • Induces cell cycle arrest
  • Can be used to design PROTAC CDK4/6 degrader

Catalog No. 50-003-39675 Supplier Medchemexpress LLC Supplier No. HY126534A50MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Abemaciclib metabolite M18 hydrochloride is a metabolite of Abemaciclib and functions as a CDK inhibitor with antitumor activity. It has been used to design PROTAC CDK4/6 degraders. This product is for research use only, suitable for laboratory chemical applications and experienced personnel. It inhibits retinoblastoma (RB) protein phosphorylation, inducing cell cycle arrest, and can increase T-cell persistence and immunologic memory.

  • Solid appearance
  • Purity of 98.18%
  • Molecular weight: 531.00
  • Chemical formula: C25H29ClF2N8O
  • Stable under recommended storage conditions
  • Not classified as a hazardous substance or mixture
  • Acts as a CDK inhibitor
  • Exhibits antitumor activity
  • Inhibits retinoblastoma (RB) protein phosphorylation
  • Induces cell cycle arrest
  • Can be used to design PROTAC CDK4/6 degrader

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.