Learn More
Medchemexpress LLC Abemaciclib metabolite M2 (LSN2839567) | 1231930-57-6 | MFCD32067988 | 99.8% | 478.54 g·mol⁻1 | C25H28F2N8 | 5 MG
SDP
Catalog No. 5000263047
Encompass_Preferred
Abemaciclib metabolite M2 is a research-grade analytical standard and a primary metabolite of the CDK4/6 inhibitor abemaciclib. It is supplied for analytical, biochemical, and preclinical research applications where an authentic metabolite reference is required. The compound shows potent CDK4 and CDK6 inhibition and is provided at high chemical purity suitable for quantitative assays.
  • Potent CDK4 inhibition (IC50 1.2 nM).
  • Potent CDK6 inhibition (IC50 1.3 nM).
  • High purity suitable for analytical work (99.75%).
  • Molecular formula C25H28F2N8; molecular weight 478.54 g·mol⁻1.
  • Intended for metabolite profiling, analytical method development, and preclinical studies.
Catalog No. 50-002-63047 Supplier Medchemexpress LLC Supplier No. HY1286695MG
Abemaciclib metabolite M2 is a research-grade analytical standard and a primary metabolite of the CDK4/6 inhibitor abemaciclib. It is supplied for analytical, biochemical, and preclinical research applications where an authentic metabolite reference is required. The compound shows potent CDK4 and CDK6 inhibition and is provided at high chemical purity suitable for quantitative assays.
  • Potent CDK4 inhibition (IC50 1.2 nM).
  • Potent CDK6 inhibition (IC50 1.3 nM).
  • High purity suitable for analytical work (99.75%).
  • Molecular formula C25H28F2N8; molecular weight 478.54 g·mol⁻1.
  • Intended for metabolite profiling, analytical method development, and preclinical studies.