Learn More
Medchemexpress LLC Abemaciclib metabolite M20 | 2138499-06-4 | 95.9% | 10 MG
SDP
Catalog No. 5000341194
Encompass_Preferred
Abemaciclib metabolite M20 (LSN3106726) is the active metabolite of Abemaciclib and functions as a selective CDK4/6 inhibitor for cancer treatment. This product is intended for research use only.
  • Induces senescence in breast cancer cells
  • Inhibits growth and cell cycle progression in non-small cell lung cancer (NSCLC) cells, colorectal cancer (CRC) cells, and breast cancer cells
  • Effective as a CDK4 and CDK6 inhibitor with an IC50 value in the range of 1-3 nM
Catalog No. 50-003-41194 Supplier Medchemexpress LLC Supplier No. HY12933610MG
Abemaciclib metabolite M20 (LSN3106726) is the active metabolite of Abemaciclib and functions as a selective CDK4/6 inhibitor for cancer treatment. This product is intended for research use only.
  • Induces senescence in breast cancer cells
  • Inhibits growth and cell cycle progression in non-small cell lung cancer (NSCLC) cells, colorectal cancer (CRC) cells, and breast cancer cells
  • Effective as a CDK4 and CDK6 inhibitor with an IC50 value in the range of 1-3 nM