Learn More
Medchemexpress LLC Abemaciclib metabolite M20 (LSN3106726) | 2138499-06-4 | 95.9% | 50 MG
SDP
Catalog No. 5000339341
Encompass_Preferred
Abemaciclib metabolite M20 is the active metabolite of Abemaciclib and acts as a selective CDK4/6 inhibitor for the treatment of cancer. It is for research use only.
  • Active metabolite of abemaciclib
  • Selective CDK4/6 inhibitor
  • Induces senescence in breast cancer cells
  • Inhibits growth and cell cycle progression of NSCLC, colorectal, and breast cancer cells
  • IC50 value for CDK4 and CDK6 in the range of 1-3 nM
Catalog No. 50-003-39341 Supplier Medchemexpress LLC Supplier No. HY12933650MG
Abemaciclib metabolite M20 is the active metabolite of Abemaciclib and acts as a selective CDK4/6 inhibitor for the treatment of cancer. It is for research use only.
  • Active metabolite of abemaciclib
  • Selective CDK4/6 inhibitor
  • Induces senescence in breast cancer cells
  • Inhibits growth and cell cycle progression of NSCLC, colorectal, and breast cancer cells
  • IC50 value for CDK4 and CDK6 in the range of 1-3 nM